MilliporeSigma
  • A carbon-carbon-coupled dimeric bergenin derivative biotransformed by Pleurotus ostreatus.
A carbon-carbon-coupled dimeric bergenin derivative biotransformed by Pleurotus ostreatus.
Bioorganic & medicinal chemistry letters (2005-07-05)
Dong Wang, Hong-Tao Zhu, Ying-Jun Zhang, Chong-Ren Yang
ABSTRACT
A novel C-C-coupled dimer derivative of bergenin was produced by the biotransformation of cultured mycelia of white rot fungus Pleurotus ostreatus. Its structure was elucidated by detailed spectroscopic analysis.
Sigma-Aldrich
Bergenin, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.