MilliporeSigma
  • The effect of selective serotonin re-uptake inhibitors on cytochrome P4502D6 (CYP2D6) activity in human liver microsomes.
The effect of selective serotonin re-uptake inhibitors on cytochrome P4502D6 (CYP2D6) activity in human liver microsomes.
British journal of clinical pharmacology (1992-09-01)
H K Crewe, M S Lennard, G T Tucker, F R Woods, R E Haddock
ABSTRACT
Inhibition of human cytochrome P4502D6 (CYP2D6)-catalysed metabolism can lead to clinically significant alterations in pharmacokinetics. Since there is evidence that the selective serotonin reuptake inhibitor (SSRI) class of antidepressant drugs might inhibit CYP2D6, the effects of five SSRIs on human liver microsomal CYP2D6 activity were compared with each other and with three tricyclic antidepressant drugs. On a molar basis, paroxetine was the most potent of the SSRIs at inhibiting the CYP2D6-catalysed oxidation of sparteine (Ki = 0.15 microM), although fluoxetine (0.60 microM) and sertaline (0.70 microM) had Ki values in the same range. Fluvoxamine (8.2 microM) and citalopram (5.1 microM) also inhibited CYP2D6 activity. The major circulating metabolites of paroxetine in man produced negligible inhibition. In contrast, norfluoxetine the active metabolite of fluoxetine, was a potent CYP2D6 inhibitor (0.43 microM). CYP2D6 activity was also diminished by the tricyclic antidepressant drugs clomipramine (2.2 microM), desipramine (2.3 microM) and amitriptyline (4.0 microM). These findings suggest that compounds with SSRI activity are likely to interact with human CYP2D6 in vivo with the potential of causing drug interactions.
Sigma-Aldrich
Thioridazine hydrochloride, ≥99%
Pricing and availability is not currently available.