MilliporeSigma
  • Repeated dosing with oral allosteric modulator of adenosine A1 receptor produces tolerance in rats with neuropathic pain.
Repeated dosing with oral allosteric modulator of adenosine A1 receptor produces tolerance in rats with neuropathic pain.
Anesthesiology (2004-04-17)
Xinhui Li, Carsten Bantel, Dawn Conklin, Steven R Childers, James C Eisenach
ABSTRACT
The positive allosteric adenosine receptor modulator, T62 (2-amino-3-(4-chlorobenzoyl)-5,6,7,8-tetrahydrobenzothiophene), has been shown to reduce mechanical allodynia in a rat model of neuropathic pain. However, whether chronic oral T62 retains efficacy in this pain model has not been examined. Therefore, the authors studied antiallodynic effects of chronic oral T62 in spinal nerved-ligated rats, as well as motor and sedative behavioral effects. Oral T62, 100 mg/kg, or oral oil was applied daily to spinal nerve-ligated rats for 4 weeks, with rat weights examined daily. Sedation, placing and ambulation scores, and withdrawal threshold were observed for 3 h daily for the first 2 weeks and then once a week. At the end of observation, the animals were killed, and the spinal tissues were collected for radioligand binding. In addition, withdrawal thresholds were also observed in rats with 5 days of treatment with 50 mg/kg oral T62. Furthermore, the effects of intrathecal adenosine on rats with oral T62 or oil treatment were compared. Chronic oral T62, at 100 mg/kg, initially returned the withdrawal threshold to mechanical testing to preinjury levels, with minor or no sedative or motor effects. Tolerance was observed, with a 60% loss of most possible effects in antiallodynia within 5 days of daily administration. Similarly, tolerance also occurred with chronic oral T62 at 50 mg/kg but did not alter the effect of intrathecal adenosine. Furthermore, 4 weeks of exposure to 100 mg/kg T62 resulted in a small reduction in spinal cord A1 receptor number. The results imply that chronically administered A1 adenosine modulators lose efficacy over time, partly as a result of receptor down-regulation.