MilliporeSigma
  • Trans-azetidine-2,4-dicarboxylic acid activates neuronal metabotropic receptors.
Trans-azetidine-2,4-dicarboxylic acid activates neuronal metabotropic receptors.
Neuroreport (1993-07-01)
M Favaron, R M Manev, P Candeo, R Arban, N Gabellini, A P Kozikowski, H Manev
ABSTRACT
The expression of metabotropic glutamate receptors (mGluRs) in primary cultures of cerebellar granule neurones can be: (i) modulated by the degree of depolarization during the culture period, rendering neurones differently sensitive to agonist-stimulated inositol phosphate (IP) hydrolysis; (ii) down-regulated by specific mGluR agonists. In this culture the new rigid glutamate analogue, (+/-)-trans-azetidine-2,4-dicarboxylic acid (t-ADA) and the known mGluR agonist 1S,3R-aminocyclopentane-1,3-dicarboxylic acid (1S,3R-ACPD) stimulated IP formation in line with the depolarization-modified expression of mGluR1. However, the two compounds caused different patterns of mGluR down-regulation. The effects of t-ADA and 1S,3R-ACPD were also tested on transformed human embryonic kidney 293 cells transfected with mGluR1. Only 1S,3R-ACPD, but not t-ADA, stimulated IP hydrolysis, suggesting that t-ADA acts on a subtype of metabotropic receptors different from mGluR1. Hence, t-ADA might prove useful in differentiating the function of various mGluR subtypes.