MilliporeSigma
  • Statins protect against fulminant pneumococcal infection and cytolysin toxicity in a mouse model of sickle cell disease.
Statins protect against fulminant pneumococcal infection and cytolysin toxicity in a mouse model of sickle cell disease.
The Journal of clinical investigation (2010-01-23)
Jason W Rosch, Angela R Boyd, Ernesto Hinojosa, Tamara Pestina, Yunming Hu, Derek A Persons, Carlos J Orihuela, Elaine I Tuomanen
ABSTRACT
Sickle cell disease (SCD) is characterized by intravascular hemolysis and inflammation coupled to a 400-fold greater incidence of invasive pneumococcal infection resulting in fulminant, lethal pneumococcal sepsis. Mechanistically, invasive infection is facilitated by a proinflammatory state that enhances receptor-mediated endocytosis of pneumococci into epithelial and endothelial cells. As statins reduce chronic inflammation, in addition to their serum cholesterol-lowering effects, we hypothesized that statin therapy might improve the outcome of pneumococcal infection in SCD. In this study, we tested this hypothesis in an experimental SCD mouse model and found that statin therapy prolonged survival following pneumococcal challenge. The protective effect resulted in part from decreased platelet-activating factor receptor expression on endothelia and epithelia, which led to reduced bacterial invasion. An additional protective effect resulted from inhibition of host cell lysis by pneumococcal cholesterol-dependent cytotoxins (CDCs), including pneumolysin. We conclude therefore that statins may be of prophylactic benefit against invasive pneumococcal disease in patients with SCD and, more broadly, in settings of bacterial pathogenesis driven by receptor-mediated endocytosis and the CDC class of toxins produced by Gram-positive invasive bacteria.