MilliporeSigma
  • Structure of the N-linked glycan present on multiple glycoproteins in the Gram-negative bacterium, Campylobacter jejuni.
Structure of the N-linked glycan present on multiple glycoproteins in the Gram-negative bacterium, Campylobacter jejuni.
The Journal of biological chemistry (2002-08-21)
N Martin Young, Jean-Robert Brisson, John Kelly, David C Watson, Luc Tessier, Patricia H Lanthier, Harold C Jarrell, Nicolas Cadotte, Frank St Michael, Erika Aberg, Christine M Szymanski
ABSTRACT
Mass spectrometry investigations of partially purified Campylobacter jejuni protein PEB3 showed it to be partially modified with an Asn-linked glycan with a mass of 1406 Da and composed of one hexose, five N-acetylhexosamines and a species of mass 228 Da, consistent with a trideoxydiacetamidohexose. By means of soybean lectin affinity chromatography, a mixture of glycoproteins was obtained from a glycine extract, and two-dimensional gel proteomics analysis led to the identification of at least 22 glycoproteins, predominantly annotated as periplasmic proteins. Glycopeptides were prepared from the glycoprotein mixture by Pronase digestion and gel filtration. The structure of the glycan was determined by using nano-NMR techniques to be GalNAc-alpha1,4-GalNAc-alpha1,4-[Glcbeta1,3-]GalNAc-alpha1,4-GalNAc-alpha1,4-GalNAc-alpha1,3-Bac-beta1,N-Asn-Xaa, where Bac is bacillosamine, 2,4-diacetamido-2,4,6-trideoxyglucopyranose. Protein glycosylation was abolished when the pglB gene was mutated, providing further evidence that the enzyme encoded by this gene is responsible for formation of the glycopeptide N-linkage. Comparison of the pgl locus with that of Neisseria meningitidis suggested that most of the homologous genes are probably involved in the biosynthesis of bacillosamine.
Sigma-Aldrich
S-Gal®/LB Agar Blend, reagent for selection of recombinant bacterial clones