MilliporeSigma
Indigestible mitochondria cause heartburn.
Cell research (2012-06-20)
Roberta A Gottlieb, Phyllis-Jean Linton
ABSTRACT
The link between impaired autophagic flux (autophagus interruptus), damaged mitochondria, and myocardial inflammation has been further tightened with the recent paper by Oka and colleagues, in which failure to degrade mitochondrial DNA exacerbated myocardial inflammation in the context of pressure overload. Using mice with cardiac-specific deletion of the lysosomal DNase II that were subjected to aortic banding, Otsu's group showed that mitochondrial DNA accumulated in lysosomes and resulted in TLR9-dependent production of inflammatory cytokines.
Sigma-Aldrich
Deoxyribonuclease II from porcine spleen, Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein