MilliporeSigma
  • Metabolic enzyme diversity in different human thyroid cell lines and their sensitivity to gravitational forces.
Metabolic enzyme diversity in different human thyroid cell lines and their sensitivity to gravitational forces.
Proteomics (2012-06-19)
Jessica Pietsch, Albert Sickmann, Gerhard Weber, Johann Bauer, Marcel Egli, Robert Wildgruber, Manfred Infanger, Daniela Grimm
ABSTRACT
Many cancer cells show unique protein expression patterns. We used proteome technology including MS, free flow isoelectric focusing and Western blotting to determine current concentrations of metabolic enzymes in healthy and malignant human thyroid cells. Three different types of human thyroid cells were investigated after they had been cultured under equal conditions. MS revealed high quantities of glycolytic enzymes and moderate quantities of citric acid cycle enzymes in malignant FTC-133 cells with abnormal LDH B-chains, high quantities of glycolytic enzymes and marginal quantities of citric acid cycle enzymes in normal HTU-5 cells, and low quantities of glycolytic enzymes and marginal quantities of citrate cycle enzymes in malignant CGTH-W1 cells with abnormal LDH A-chains. When an alteration of gene expression activity was challenged physically by removing gravity forces, the concentrations of various glycolytic enzymes were changed in normal and malignant thyroid cells. However, the changes varied among the different cell types. Different cellular alignment of the enzymes could be one reason for the cell type-specific behavior as demonstrated by histological analysis of alpha-enolase.
Sigma-Aldrich
Triosephosphate Isomerase from baker′s yeast (S. cerevisiae), Type I, ammonium sulfate suspension, ~10,000 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type X, lyophilized powder, ≥3,500 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type III-S, ammonium sulfate suspension, ≥4,000 units/mg protein