MilliporeSigma
  • Mu opioid receptor phosphorylation, desensitization, and ligand efficacy.
Mu opioid receptor phosphorylation, desensitization, and ligand efficacy.
The Journal of biological chemistry (1997-11-20)
Y Yu, L Zhang, X Yin, H Sun, G R Uhl, J B Wang
ABSTRACT
Mu opioid receptors are subject to phosphorylation and desensitization through actions of at least two distinct biochemical pathways: agonist-dependent mu receptor phosphorylation and desensitization induced by a biochemically distinct second pathway dependent on protein kinase C activation (1). To better understand the nature of the agonist-induced mu receptor phosphorylation events, we have investigated the effects of a variety of opiate ligands of varying potencies and intrinsic activities on mu receptor phosphorylation and desensitization. Exposure to the potent full agonists sufentanil, dihydroetorphine, etorphine, etonitazine, and [D-Ala2, MePhe4, Glyol5]enkephalin (DAMGO) led to strong receptor phosphorylation, while methadone, l-alpha-acetylmethadone (LAAM), morphine, meperidine, DADL, beta-endorphin(1-31), enkephalins, and dynorphin A(1-17) produced intermediate effects. The partial agonist buprenorphine minimally enhanced receptor phosphorylation while antagonists failed to alter phosphorylation. Buprenorphine and full antagonists each antagonized the enhanced mu receptor phosphorylation induced by morphine or DAMGO. The rank order of opiate ligand efficacies in producing mu receptor-mediated functional desensitization generally paralleled their rank order of efficacies in producing receptor phosphorylation. Interestingly, the desensitization and phosphorylation mediated by methadone and LAAM were disproportionate to their efficacies in two distinct test systems. This generally good fit between the efficacies of opiates in mu receptor activation, phosphorylation, and desensitization supports the idea that activated receptor/agonist/G-protein complexes and/or receptor conformational changes induced by agonists are required for agonist-induced mu receptor phosphorylation. Data for methadone and LAAM suggest possible contribution from their enhanced desensitizing abilities to their therapeutic efficacies.
Sigma-Aldrich
(±)-Methadone hydrochloride, powder, ≥98%