MilliporeSigma
  • Construction of an expression system for the secretory production of recombinant α-agarase in yeast.
Construction of an expression system for the secretory production of recombinant α-agarase in yeast.
Biotechnology letters (2012-02-09)
Ji-Hwan Seok, Hye-Soo Kim, Yuji Hatada, Soo-Wan Nam, Yeon-Hee Kim
ABSTRACT
α-Agarase hydrolyzes the α-1,3 linkage of agarose yielding agaro-oligosaccharides. It is less well characterized than β-agarase. AgaA gene (2.3 kb ORF), encoding the α-agarase from Thalassomonas JAMB A33, was subcloned into both a constitutive and an inducible expression vector. Both the constructed plasmids, pVT-AgaA (ADH1 promoter) and pYInu-AgaA (GAL10 promoter), were transformed into Saccharomyces cerevisiae SEY2102 and FY833 and pPIC9-AgaA harboring the AOX1 promoter was transformed into Pichia pastoris GS115. The recombinant α-agarases were over-expressed with activities from 0.3 to 1.6 unit/ml, the highest being in the SEY2102/pYInu-AgaA transformant. Most of the recombinant α-agarase was extracellular because each plasmid possesses a signal sequence for the secretory production of α-agarase. In contrast, the Pichia host-vector expression system was unsuitable for the production of recombinant α-agarase. This is the first report of recombinant production of α-agarase in yeast for industrial use.
Sigma-Aldrich
Agarase from Pseudomonas atlantica, lyophilized powder, ≥5,000 units/mg protein (Lowry)
Pricing and availability is not currently available.