MilliporeSigma
  • Improvement in the catalytic activity of β-agarase AgaA from Zobellia galactanivorans by site-directed mutagenesis.
Improvement in the catalytic activity of β-agarase AgaA from Zobellia galactanivorans by site-directed mutagenesis.
Journal of microbiology and biotechnology (2011-12-01)
Seungwoo Lee, Dong-Geun Lee, Min-Kyung Jang, Myong-Je Jeon, Hye-Ji Jang, Sang-Hyeon Lee
ABSTRACT
In this study, site-directed mutagenesis was performed on the β-agarase AgaA gene from Zobellia galactanivorans to improve its catalytic activity and thermostability. The activities of three mutant enzymes, S63K, C253I, and S63K-C253I, were 126% (1,757.78 U/mg), 2.4% (33.47 U/mg), and 0.57% (8.01 U/mg), respectively, relative to the wildtype beta-agarase AgaA (1,392.61 U/mg) at 40°C. The stability of the mutant S63K enzyme was 125% of the wild-type up to 45°C, where agar is in a sol state. The mutant S63K enzyme produced 166%, 257%, and 220% more neoagarohexaose, and 230%, 427%, and 350% more neoagarotetraose than the wild-type in sol, gel, and nonmelted powder agar, respectively, at 45°C over 24 h. The mutant S63K enzyme produced 50% more neoagarooligosaccharides from agar than the wild-type beta-agarase AgaA from agarose under the same conditions. Thus, mutant S63K β-agarase AgaA may be useful for the production of functional neoagarooligosaccharides.
Sigma-Aldrich
Agarase from Pseudomonas atlantica, lyophilized powder, ≥5,000 units/mg protein (Lowry)
Pricing and availability is not currently available.