MilliporeSigma
  • A role for inositol monophosphatase 1 (IMPA1) in salinity adaptation in the euryhaline eel (Anguilla anguilla).
A role for inositol monophosphatase 1 (IMPA1) in salinity adaptation in the euryhaline eel (Anguilla anguilla).
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2010-06-16)
Svetlana Kalujnaia, Jill McVee, Taciana Kasciukovic, Alan J Stewart, Gordon Cramb
ABSTRACT
This study investigated the expression and tissue distribution of inositol monophosphatase (IMPA1) and characterized its role in salinity adaptation in the eel. The coding sequence of eel IMPA1 was determined and confirmed to be orthologous to the mammalian gene/enzyme by phylogenetic analysis and structural modeling. Quantitative real-time PCR and Western blot techniques indicated up to 17-fold increases in mRNA expression and 2-fold increases in protein abundance in major osmoregulatory tissues following transfer of fish to seawater (SW). This was accompanied by up to 5-fold increases in enzyme activity, and 1.8- and 3-fold increases in inositol contents within the gill and kidney, respectively. Immunohistological studies revealed that IMPA1 protein expression predominated in SW-acclimated fish within basal epithelial/epidermal layers of the gill, esophagus, intestine, skin, and fins. SW transfer also induced a 10-fold increase in inositol content in the fin. IMPA1 immunoreactivity was also identified in chondrocytes within the cartilagenous matrix of the gills and fins, as well as in clusters of interstitial cells surrounding the kidney tubules. The observed increases in expression of IMPA1 highlight a protective role for inositol within various eel tissues following SW acclimation. This constitutes an adaptive mechanism in teleost fish naturally exposed to hypertonic environments.