MilliporeSigma
  • Involvement of prostaglandin F2alpha in the adverse effect of PCB 77 on the force of contractions of bovine myometrium.
Involvement of prostaglandin F2alpha in the adverse effect of PCB 77 on the force of contractions of bovine myometrium.
Toxicology (2009-06-25)
Michal H Wrobel, Robert Rekawiecki, Jan Kotwica
ABSTRACT
Polychlorinated biphenyls (PCBs) stimulate in vitro both the force of myometrial contractions and endometrial secretion of PGF2alpha in cattle. Therefore, the goal of this study was to investigate the participation of PGF2alpha in the effect of PCBs on uterine contractility. For this aim, the myometrial strips were incubated (48h) with PCB 77 at the dose of 1, 10 and 100ng/ml (i.e., 0.0034, 0.034 and 0.34nmol/ml) separately or jointly with indomethacin (INDO, 10(-4)M), which blocks the PGF2alpha synthesis. Next, the force of myometrial strips contractions was measured. Further, the influence of PCB 77 (0.1, 1 and 10ng/ml) on the PGF2alpha secretion from myometrial cells after 6, 24, and 48h and PCB 77 (1 and 10ng/ml) on the mRNA expression of cyclooxygenase 2 (COX-2) and PGF2alpha synthase (PGFS) in myometrial cells after 6 and 24h, was investigated. The increase (P<0.05-0.001) of the contractions force of myometrial strips evoked by each dose of PCB 77, was markedly reduced (P<0.05-001) by INDO. There was an increase (P<0.05-0.001) of both PGF2alpha secretion after all studied periods of cell incubation and mRNA expression for COX-2 and PGFS after 6h treatment of myometrial cells with PCB 77. It can be concluded that myometrial synthesis of PGF2alpha and its further secretion is a part of the mechanism by means of which PCB 77 may affect the force of myometrial contractions in cattle.
Sigma-Aldrich
Prostaglandin F Synthase human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)