MilliporeSigma
  • A novel endoglycoceramidase hydrolyzes oligogalactosylceramides to produce galactooligosaccharides and ceramides.
A novel endoglycoceramidase hydrolyzes oligogalactosylceramides to produce galactooligosaccharides and ceramides.
The Journal of biological chemistry (2007-01-25)
Yohei Ishibashi, Toru Nakasone, Masashi Kiyohara, Yasuhiro Horibata, Keishi Sakaguchi, Atsushi Hijikata, Sachiyo Ichinose, Akira Omori, Yasuyuki Yasui, Akihiro Imamura, Hideharu Ishida, Makoto Kiso, Nozomu Okino, Makoto Ito
ABSTRACT
Enzymes capable of hydrolyzing the beta-glycosidic linkage between oligosaccharides and ceramides in various glycosphingolipids has been found in microorganisms and invertebrates and designated endoglycoceramidase (EC 3.2.1.123) or ceramide glycanase. Here we report the molecular cloning, characterization, and homology modeling of a novel endoglycoceramidase that hydrolyzes oligogalactosylceramides to produce galactooligosaccharides and ceramides. The novel enzyme was purified from a culture supernatant of Rhodococcus equi, and the gene encoding 488 deduced amino acids was cloned using peptide sequences of the purified enzyme. Eight residues essential for the catalytic reaction in microbial and animal endoglycoceramidases were all conserved in the deduced amino acid sequence of the novel enzyme. Homology modeling of the enzyme using endocellulase E1 as a template revealed that the enzyme displays a (beta/alpha)8 barrel structure in which Glu234 at the end of beta-strand 4 and Glu341 at the end of beta-strand 7 could function as an acid/base catalyst and a nucleophile, respectively. Site-directed mutagenesis of these glutamates resulted in a complete loss of the activity without a change in their CD spectra. The recombinant enzyme hydrolyzed the beta-galactosidic linkage between oligosaccharides and ceramides of 6-gala series glycosphingolipids that were completely resistant to hydrolysis by the enzymes reported so far. In contrast, the novel enzyme did not hydrolyze ganglio-, globo-, or lactoseries glycosphingolipids. The enzyme is therefore systematically named "oligogalactosyl-N-acylsphingosine 1,1'-beta-galactohydrolase" or tentatively designated "endogalactosylceramidase."
Sigma-Aldrich
Endoglycoceramidase II from Rhodococcus sp., aqueous solution
Pricing and availability is not currently available.