MilliporeSigma
  • Determination of low molecular weight organic acids in soil, plants, and water by capillary zone electrophoresis.
Determination of low molecular weight organic acids in soil, plants, and water by capillary zone electrophoresis.
Analytical and bioanalytical chemistry (2003-03-29)
Ying-Hui Li, Bi-Xia Huang, Xiao-Quan Shan
ABSTRACT
Determination of low molecular weight organic acids in soils and plants by capillary zone electrophoresis was accomplished using a phthalate buffer and indirect UV detection mode. The influence of some crucial parameters, such as pH, buffer concentration and surfactant were investigated. A good separation of seven organic acids was achieved within 5 min using an electrolyte containing 15 mmol L(-1) potassium hydrogen phthalate, 0.5 mmol L(-1) myristyltrimethylammonium bromide (MTAB), and 5% methanol (MeOH) (v/v) at pH 5.60, separation voltage -20 kV, and temperature 25 degrees C. The relative standard deviation (n=5) of the method was found to be in range 0.18-0.56% for migration time and 3.2-4.8% for peak area. The limit of detection ranged between 0.5 micro mol L(-1) to 6 micro mol L(-1) at a signal-to-noise ratio of 3. The recovery of standard organic acids added to real samples ranged from 87 to 119%. This method was simple, rapid and reproducible, and could be applied to the simultaneous determination of organic acids in environmental samples.
Sigma-Aldrich
Myristyltrimethylammonium bromide, ≥99%
Pricing and availability is not currently available.