MilliporeSigma
  • Antibody-induced internalization of viral glycoproteins in pseudorabies virus-infected monocytes and role of the cytoskeleton: a confocal study.
Antibody-induced internalization of viral glycoproteins in pseudorabies virus-infected monocytes and role of the cytoskeleton: a confocal study.
Veterinary microbiology (2002-03-13)
Gerlinde R Van de Walle, Herman W Favoreel, Hans J Nauwynck, Patrick Van Oostveldt, Maurice B Pensaert
ABSTRACT
Addition of pseudorabies virus (PrV)-specific polyclonal immunoglobulins to PrV-infected monocytes induces internalization of plasma membrane anchored viral glycoproteins. This process may interfere with antibody-dependent cell lysis and resembles the well-studied physiological endocytosis process. A confocal study was designed to investigate whether the major cellular components, involved in physiological endocytosis (clathrin, actin, dynein and microtubules), play a role in this virological internalization process. In order to visualize the interaction of endosomes, which contain the internalized viral glycoproteins, with clathrin, actin, dynein and microtubules, a double labeling of viral glycoproteins and different cellular proteins was performed. Porcine monocytes were inoculated with the PrV-strain 89V87 at a multiplicity of infection of 50 for 13h. After the addition of FITC-labeled porcine polyclonal PrV-specific antibodies, cells were fixed with para-formaldehyde at different time points and afterwards permeabilized. The different cellular components were visualized with monoclonal antibodies and a Texas Red-conjugate, with the exception of actin, which was stained with phalloidin-Texas Red. The cells were analyzed by confocal microscopy. A clear co-localization was observed between the viral glycoproteins and clathrin and dynein during the internalization process. The microtubules were in close contact with the internalized vesicles. For actin no co-localization could be observed. It can be stated that clathrin, dynein and microtubules, important components during physiological endocytosis, are also of importance during the antibody-induced internalization of viral glycoproteins.
Sigma-Aldrich
Phalloidin–Atto 655, suitable for fluorescence, ≥90.0% (sum of isomers, HPLC)