MilliporeSigma
  • Cell wall composition of tomato fruit changes during development and inhibition of vesicle trafficking is associated with reduced pectin levels and reduced softening.
Cell wall composition of tomato fruit changes during development and inhibition of vesicle trafficking is associated with reduced pectin levels and reduced softening.
Plant physiology and biochemistry : PPB (2013-03-19)
Daniel Lunn, Thanh D Phan, Gregory A Tucker, Grantley W Lycett
ABSTRACT
Fruit development entails a multitude of biochemical changes leading up to the mature green stage. During this period the cell wall will undergo complex compositional and structural changes. Inhibition of genes encoding elements of the machinery involved in trafficking to the cell wall presents us with a useful tool to study these changes and their associated phenotypes. An antisense SlRab11a transgene has previously been shown to reduce ripening-associated fruit softening. SlRab11a is highly expressed during fruit development which is associated with a period of pectin influx into the wall. We have analysed the cell wall polysaccharides at different stages of growth and ripening of wild type and antisense SlRab11a transgenic tomato (Solanum lycopersicum cv, Ailsa Craig) fruit. Our results demonstrated intriguing changes in cell wall composition during the development and ripening of wild type Alisa Craig tomato fruit. Analysis of SlRab11a expression by TaqMan PCR showed it to be expressed most strongly during growth of the fruit, suggesting a possible role in cell wall deposition. The SlRab11a antisense fruit had a decreased proportion of pectin in the cell wall compared with the wild type. We suggest a new approach for modification of fruit shelf-life by changing cell wall deposition rather than cell wall hydrolytic enzymes.
Sigma-Aldrich
GenElute Plant Genomic DNA Miniprep Kit, sufficient for 350 purifications