MilliporeSigma
  • Substrate elasticity provides mechanical signals for the expansion of hemopoietic stem and progenitor cells.
Substrate elasticity provides mechanical signals for the expansion of hemopoietic stem and progenitor cells.
Nature biotechnology (2010-10-05)
Jeff Holst, Sarah Watson, Megan S Lord, Steven S Eamegdool, Daniel V Bax, Lisa B Nivison-Smith, Alexey Kondyurin, Liang Ma, Andres F Oberhauser, Anthony S Weiss, John E J Rasko
ABSTRACT
Surprisingly little is known about the effects of the physical microenvironment on hemopoietic stem and progenitor cells. To explore the physical effects of matrix elasticity on well-characterized primitive hemopoietic cells, we made use of a uniquely elastic biomaterial, tropoelastin. Culturing mouse or human hemopoietic cells on a tropoelastin substrate led to a two- to threefold expansion of undifferentiated cells, including progenitors and mouse stem cells. Treatment with cytokines in the presence of tropoelastin had an additive effect on this expansion. These biological effects required substrate elasticity, as neither truncated nor cross-linked tropoelastin reproduced the phenomenon, and inhibition of mechanotransduction abrogated the effects. Our data suggest that substrate elasticity and tensegrity are important mechanisms influencing hemopoietic stem and progenitor cell subsets and could be exploited to facilitate cell culture.
Sigma-Aldrich
Tropoelastin human, recombinant, expressed in E. coli, 1 mg
Pricing and availability is not currently available.