MilliporeSigma
  • Treatment of thin stillage in a high-rate anaerobic fluidized bed bioreactor (AFBR).
Treatment of thin stillage in a high-rate anaerobic fluidized bed bioreactor (AFBR).
Bioresource technology (2012-08-07)
Mehran Andalib, Hisham Hafez, Elsayed Elbeshbishy, George Nakhla, Jesse Zhu
ABSTRACT
The primary objective of this work was to investigate the treatability of thin stillage as a by-product of bioethanol production plants using an anaerobic fluidized bed bioreactor (AFBR) employing zeolite with average diameter of (d(m)) of 425-610 μm and specific surface area (SSA) of 26.5m(2)/g as the carrier media. Despite the very high strength of thin stillage with chemical oxygen demand of 130,000 mg TCOD/L and suspended solids of 47,000 mg TSS/L, the AFBR showed up to 88% TCOD and 78% TSS removal at very high organic and solids loading rates (OLR and SLR) of 29 kg COD/m(3)d and 10.5 kg TSS/m(3)d respectively and hydraulic retention time (HRT) of 3.5 days. Methane production rates of up to 160 L/d at the steady state equivalent to 40 L(CH4)/L(thin stillage)d and biogas production rate per reactor volume of 15.8L(gas)/L(reactor)d were achieved.
Sigma-Aldrich
Zeolite