MilliporeSigma
  • Antifungal activity of lariciresinol derived from Sambucus williamsii and their membrane-active mechanisms in Candida albicans.
Antifungal activity of lariciresinol derived from Sambucus williamsii and their membrane-active mechanisms in Candida albicans.
Biochemical and biophysical research communications (2011-06-18)
Bomi Hwang, Jaeyong Cho, In-sok Hwang, Hong-Guang Jin, Eun-Rhan Woo, Dong Gun Lee
ABSTRACT
Lariciresinol is an enterolignan precursor isolated from the herb Sambucus williamsii, a folk medicinal plant used for its therapeutic properties. In this study, the antifungal properties and mode of action of lariciresinol were investigated. Lariciresinol displays potent antifungal properties against several human pathogenic fungal strains without hemolytic effects on human erythrocytes. To understand the antifungal mechanism of action of lariciresinol, the membrane interactions of lariciresinol were examined. Fluorescence analysis using the membrane probe 3,3'-diethylthio-dicarbocyanine iodide (DiSC(3)-5) and 1,6-diphenyl-1,3,5-hexatriene (DPH), as well as a flow cytometric analysis with propidium iodide (PI), a membrane-impermeable dye, indicated that lariciresinol was associated with lipid bilayers and induced membrane permeabilization. Therefore, the present study suggests that lariciresinol possesses fungicidal activities by disrupting the fungal plasma membrane and therapeutic potential as a novel antifungal agent for the treatment of fungal infectious diseases in humans.
Sigma-Aldrich
(+)-Lariciresinol, ≥95.0% (HPLC)