MilliporeSigma
  • Liposomal gD ectodomain (gD1-306) vaccine protects against HSV2 genital or rectal infection of female and male mice.
Liposomal gD ectodomain (gD1-306) vaccine protects against HSV2 genital or rectal infection of female and male mice.
Vaccine (2009-10-20)
K Olson, P Macias, S Hutton, W A Ernst, G Fujii, J P Adler-Moore
ABSTRACT
Herpes simplex virus type 2 (HSV2) is the most common causative agent of genital herpes, with infection rates as high as 1 in 6 adults. The present studies were done to evaluate the efficacy of a liposomal HSV2 gD(1-306) vaccine (L-gD(1-306)-HD) in an acute murine HSV2 infection model of intravaginal (female) or intrarectal (male or female) challenge. Two doses of L-gD(1-306)-HD containing 60 microg gD(1-306)-HD and 15 microg monophosphoryl lipid A (MPL) per dose provided protection against HSV2 intravaginal challenge (86-100% survival, P< or =0.0003 vs. control liposomes; P=0.06 vs. L-gD(1-306)-HD without MPL). Both male and female mice (BALB/c and C57BL/6) immunized with L-gD(1-306)-HD/MPL were significantly protected against HSV2 intrarectal challenge, with higher survival rates compared to controls (71-100%, P< or =0.007). L-gD(1-306)-HD/MPL also provided increased survival when compared to a liposomal peptide vaccine, L-gD(264-285)-HD/MPL (male BALB/c, P</=0.001; female BALB/c and male C57BL/6, P=0.06). Mice given L-gD(1-306)-HD/MPL also had minimal disease signs, reduced viral burden in their spinal cords and elevated neutralizing antibody titers in the females. The vaccine also stimulated gD(1-306)-HD specific splenocytes of both male and female mice with significantly elevated levels of IFN-gamma compared to IL-4 (P< or =0.01) indicating that there was an enhanced Th1 response. These results provide the first evidence that the L-gD(1-306)-HD vaccine can protect both male and female mice against intrarectal HSV2 challenge.
Sigma-Aldrich
Lipid A, monophosphoryl from Escherichia coli F583 (Rd mutant), lyophilized powder