Merck
  • Chemical induction of splice-neoantigens attenuates tumor growth in a preclinical model of colorectal cancer.
Chemical induction of splice-neoantigens attenuates tumor growth in a preclinical model of colorectal cancer.
Science translational medicine (2022-12-01)
Shingo Matsushima, Masahiko Ajiro, Kei Iida, Kenji Chamoto, Tasuku Honjo, Masatoshi Hagiwara
ABSTRACT
Neoantigen production is a determinant of cancer immunotherapy. However, the expansion of neoantigen abundance for cancer therapeutics is technically challenging. Here, we report that the synthetic compound RECTAS can induce the production of splice-neoantigens that could be used to boost antitumor immune responses. RECTAS suppressed tumor growth in a CD8+ T cell- and tumor major histocompatibility complex class I-dependent manner and enhanced immune checkpoint blockade efficacy. Subsequent transcriptome analysis and validation for immunogenicity identified six splice-neoantigen candidates whose expression was induced by RECTAS treatment. Vaccination of the identified neoepitopes elicited T cell responses capable of killing cancer cells in vitro, in addition to suppression of tumor growth in vivo upon sensitization with RECTAS. Collectively, these results provide support for the further development of splice variant-inducing treatments for cancer immunotherapy.
Product Description
RECTAS, ≥98% (HPLC)