MilliporeSigma
  • Research on Application of Japanese Quince (Chaenomeles L.) and Pork Collagen in Dark Chocolate-Benefits in Prevention of Inflammation In Vitro Model.
Research on Application of Japanese Quince (Chaenomeles L.) and Pork Collagen in Dark Chocolate-Benefits in Prevention of Inflammation In Vitro Model.
Nutrients (2024-06-19)
Szymon Byczkiewicz, Dominik Szwajgier, Ewa Baranowska-Wójcik, Aleksandra Telichowska, Krystyna Szymandera-Buszka, Janusz Wojtczak, Joanna Kobus-Cisowska
ABSTRACT
In the present study, the effect of the addition of quince and collagen type I and III to dessert chocolate on its functional properties was determined. The study evaluated the antioxidant potential of the tested formulations using the FRAP method and the linoleic acid oxidation test and beta-carotene bleaching test. The tested samples were also evaluated for inhibitory activity against enzymes important in preventive health (inflammation and neurodegenerative disorders) namely: AChE, BChE, GR, GPx, COX, and SOD. The addition of quince and collagen to the chocolate samples resulted in higher activity compared to the control sample, as indicated by the FRAP test. The experiment highlighted the impact of including quince fruit on the antioxidant activity of the chocolate samples. Interestingly, merely increasing the quince fruit amount did not consistently enhance antioxidant potential. Specifically, chocolate samples with a lower proportion of quince fruit (2 g/100 g) exhibited greater antioxidant activity when supplemented with collagen I. Conversely, in samples with higher quince percentages (3 g and 4 g), those enriched with collagen III showed higher antioxidant activity. Similar correlations were observed in the linoleic acid oxidation test. Notably, samples containing 3 g and 4 g of quince and type III collagen demonstrated statistically similar highest antioxidant properties. Regardless of the collagen type used, there was no observed increase in activity towards the tested enzymes for samples with the lowest percentage of quince fruit. Both collagen types exhibited the highest activity in the inhibition assay against acetylcholinesterase and butyrylcholinesterase when combined with 3 g and 4 g of quince. Overall, the experimental incorporation of both fruit and collagen enhanced the chocolates' activity. Similarly to the antioxidant activity findings, chocolates with lower quince fruit quantities showed increased activity when supplemented with collagen III, while those with higher quince content (3 g and 4 g) displayed higher activity with collagen I. Bitter chocolate by itself is an attractive food product, rich in many bioactive compounds. However, enriching it with other attractive raw materials can make its properties and taste even more attractive.
Sigma-Aldrich
Xanthine Oxidase from bovine milk, Grade IV, ammonium sulfate suspension, ≥0.1 units/mg protein