MilliporeSigma
  • Effect of trypsin inhibitor activity in soya bean on growth performance, protein digestibility and incidence of sub-clinical necrotic enteritis in broiler chicken flocks.
Effect of trypsin inhibitor activity in soya bean on growth performance, protein digestibility and incidence of sub-clinical necrotic enteritis in broiler chicken flocks.
British poultry science (2011-07-08)
M W C D Palliyeguru, S P Rose, A M Mackenzie
ABSTRACT
1. The effect of three different levels of dietary trypsin inhibitor activity (achieved by varying the amount of non-toasted full fat soya bean in replacement for toasted full fat soya bean) on the incidence of spontaneously-occurring sub-clinical necrotic enteritis (NE) in broiler chickens was compared. A fourth dietary treatment compared the effect of a diet that used potato protein concentrate as the major protein source. The determined trypsin inhibitor activity increased with the increasing content of non-toasted soya bean: 1·90, 6·21, 8·46 and 3·72 mg/g for the three soya bean diets (0, 100 and 200 g of non-toasted soya bean/kg) and the potato protein diet respectively. 2. Although increasing amounts of the non-toasted full-fat soya bean increased the feed intakes of the birds, there was a marked reduction in protein digestibility, weight gain and feed conversion efficiency. 3. There was a linear increase in sub-clinical NE lesions in the duodenum, jejunum, mid small intestine and ileum with increasing non-toasted soya bean. Caecal Clostridium perfringens counts increased with the increasing dietary content of non-toasted soya bean. Serum α-toxin antibodies were higher in the birds fed the 200 g non-toasted soya bean/kg diet compared with the other diets. 4. The results demonstrated that variation in the amount of non-toasted dietary soya bean not only affects growth performance of broilers but also affects the incidence of sub-clinical necrotic enteritis in the flock. Ensuring the lowest possible trypsin-inhibitor activity in soya bean samples is a valuable tool to improve the health and welfare of birds and in reducing the financial losses from this disease.
Sigma-Aldrich
Anti-Chicken IgY (IgG) (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution