MilliporeSigma
  • Arginine methylation of caspase-8 controls life/death decisions in extrinsic apoptotic networks.
Arginine methylation of caspase-8 controls life/death decisions in extrinsic apoptotic networks.
Oncogene (2024-05-11)
Fabian Wohlfromm, Nikita V Ivanisenko, Sabine Pietkiewicz, Corinna König, Kamil Seyrek, Thilo Kähne, Inna N Lavrik
ABSTRACT
Procaspase-8 is a key mediator of death receptor (DR)-mediated pathways. Recently, the role of post-translational modifications (PTMs) of procaspase-8 in controlling cell death has received increasing attention. Here, using mass spectrometry screening, pharmacological inhibition and biochemical assays, we show that procaspase-8 can be targeted by the PRMT5/RIOK1/WD45 methylosome complex. Furthermore, two potential methylation sites of PRMT5 on procaspase-8, R233 and R435, were identified in silico. R233 and R435 are highly conserved in mammals and their point mutations are among the most common mutations of caspase-8 in cancer. The introduction of mutations at these positions resulted in inhibitory effects on CD95L-induced caspase-8 activity, effector caspase activation and apoptosis. In addition, we show that procaspase-8 can undergo symmetric di-methylation. Finally, the pharmacological inhibition of PRMT5 resulted in the inhibitory effects on caspase activity and apoptotic cell death. Taken together, we have unraveled the additional control checkpoint in procaspase-8 activation and the arginine methylation network in the extrinsic apoptosis pathway.
Sigma-Aldrich
Protein Methyltransferase Inhibitor, AMI-5, The Protein Methyltransferase Inhibitor, AMI-5, also referenced under CAS 17372-87-1, controls the biological activity of Protein Methyltransferase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.