MilliporeSigma
  • Integrin αvβ6 mediates the immune escape through regulation of PD-L1 and serves as a novel marker for immunotherapy of colon carcinoma.
Integrin αvβ6 mediates the immune escape through regulation of PD-L1 and serves as a novel marker for immunotherapy of colon carcinoma.
American journal of cancer research (2024-06-11)
Jintao Yu, Tianyu E, Mingliang Zhou, Jun Niu, Jinshen Wang, Ruizheng Miao, Cunjin Dong, Huijie Gao, Changqing Jing, Benjia Liang
ABSTRACT
The immune escape of colon cancer and its role in the response to immunotherapies such as PD-1/PD-L1 checkpoint inhibitors have long been of great interest. The positive outcomes of immunotherapy are limited by the immunosuppressive nature of the tumor microenvironment. Integrin αvβ6, which can regulate the progression of colon cancer, was recently reported to be involved in the immune suppression of colon cancer. In the present study, we explored the correlation between αvβ6 and PD-L1 expression by immunohistochemistry of colon cancer tissues. Then, the regulation of PD-L1 signaling by αvβ6 in colon cancer cells was demonstrated. We constructed an in vivo model and performed immunophenotyping experiments to analyze further the regulation of the immune response by αvβ6. The role of αvβ6 in the response to anti-PD-1 therapy in colon cancer was also verified. αvβ6-positive tissues exhibited increased PD-L1 expression. Inhibition of αvβ6 not only downregulated constitutive PD-L1 expression but also decreased IFN-γ-induced PD-L1 expression. In addition, αvβ6-induced PD-L1 expression was suppressed by the ERK inhibitor PD98059, and knockdown of the β6-ERK2 binding site had the equivalent effect. αvβ6 decreased CD8+ T cell infiltration and granzyme B expression in CD8+ T cells in colon cancer patients. Furthermore, mice engrafted with αvβ6-expressing colon cancer cells exhibited an unsatisfactory response to anti-PD-1 therapy, and anti-PD-1-induced increases in CD4+ and CD8+ T cell infiltration could be inhibited by αvβ6. These results indicate that αvβ6 mediates immune escape in colon cancer by upregulating PD-L1 through the ERK/MAPK pathway. Moreover, αvβ6 could serve as a marker for the efficacy of anti-PD-1 therapy in colon cancer.
Sigma-Aldrich
Anti-ITGB6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution