MilliporeSigma
Species diversity of Trichoderma in Poland.
Journal of applied genetics (2011-04-06)
Lidia Błaszczyk, Delfina Popiel, Jerzy Chełkowski, Grzegorz Koczyk, Gary J Samuels, Krzysztof Sobieralski, Marek Siwulski
ABSTRACT
In the present study, we reinvestigate the diversity of Trichoderma in Poland utilizing a combination of morphological and molecular/phylogenetic methods. A total of 170 isolates were collected from six different substrata at 49 sites in Poland. These were divided among 14 taxa as follows: 110 of 170 Trichoderma isolates were identified to the species level by the analysis of their ITS1, ITS2 rDNA sequences as: T. harzianum (43 isolates), T. aggressivum (35), T. citrinoviride (11), T. hamatum (9), T. virens (6), T. longibrachiatum (4), T. polysporum (1), and T. tomentosum (1); 60 isolates belonging to the Viride clade were identified based on a fragment of the translation-elongation factor 1-alpha (tef1) gene as: T. atroviride (20 isolates), T. gamsii (2), T. koningii (17), T. viridescens (13), T. viride (7), and T. koningiopsis (1). Identifications were made using the BLAST interface in TrichOKEY and TrichoBLAST ( http://www.isth.info ). The most diverse substrata were soil (nine species per 22 isolates) and decaying wood (nine species per 75 isolates). The most abundant species (25%) isolated from all substrata was T. harzianum.
Product Description
Millipore
Czapek-Dox broth, suitable for microbiology, NutriSelect® Basic
Pricing and availability is not currently available.