MilliporeSigma
  • Oxytocin Receptors on Calvarial Periosteal Innervation: Therapeutic Target for Post-Traumatic Headache?
Oxytocin Receptors on Calvarial Periosteal Innervation: Therapeutic Target for Post-Traumatic Headache?
Pharmaceutics (2024-06-27)
Vimala N Bharadwaj, Michael Klukinov, Robert Paul Cowan, Nazanin Mahinparvar, David John Clark, David Clifford Yeomans
ABSTRACT
Following a mild traumatic brain injury (mTBI), the most prevalent and profoundly debilitating occurrence is the emergence of an acute and persistent post-traumatic headache (PTH), for which there are presently no approved treatments. A crucial gap in knowledge exists regarding the consequences of an mTBI, which could serve as a foundation for the development of therapeutic approaches. The activation of trigeminal sensory nerve terminals that innervate the calvarial periosteum (CP)-a densely innervated tissue layer covering the calvarial skull-has been implicated in both migraines and PTHs. We have previously shown that trigeminal oxytocin receptors (OTRs) may provide a therapeutic target for PTHs. This study examined the expression of oxytocin receptors on trigeminal nerves innervating the periosteum and whether these receptors might serve as a therapeutic target for PTHs using a direct application of oxytocin to the periosteum in a rodent model of PTH. We used retrograde tracing and immunohistochemistry to determine if trigeminal ganglion (TG) neurons innervating the periosteum expressed OTRs and/or CGRPs. To model the impact of local inflammation that occurs following an mTBI, we applied chemical inflammatory mediators directly to the CP and assessed for changes in immediate-early gene expression as an indication of neuronal activation. We also determined whether mTBI would lead to expression changes to OTR levels. To determine whether these OTRs could be a viable therapeutic target, we assessed the impact of oxytocin injections into the CP in a mouse model of PTH-induced periorbital allodynia. The results of these experiments demonstrate the following: (1) the cell bodies of CP afferents reside in the TG and express both OTRs and CGRPs; (2) inflammatory chemical stimulation of the periosteum leads to rapid activation of TG neurons (phospho-ERK (p-ERK) expression), (3) mTBI-induced inflammation increased OTR expression compared to the sham group; and (4) administration of oxytocin into the periosteum on day 2 and day 40 blocked cutaneous allodynia for up to one hour post-administration for both acute and persistence phases in the PTH model-an effect that was preventable by the administration of an OTR antagonist. Taken together, our observations suggest that periosteal trigeminal afferents contribute to post-TBI craniofacial pain, and that periosteum tissue can be used as a potential local target for therapeutics such as oxytocin.
Sigma-Aldrich
L-368,899, ≥98% (HPLC), powder