MilliporeSigma
  • DMXL2 Is Required for Endocytosis and Recycling of Synaptic Vesicles in Auditory Hair Cells.
DMXL2 Is Required for Endocytosis and Recycling of Synaptic Vesicles in Auditory Hair Cells.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2024-08-16)
Hu Peng, Longhao Wang, Yunge Gao, Huihui Liu, Guotong Lin, Yu Kong, Pengcheng Xu, Hongchao Liu, Qingyue Yuan, Huanhai Liu, Lei Song, Tao Yang, Hao Wu
ABSTRACT
Ribbon synapses of inner hair cells (IHCs) are uniquely designed for ultrafast and indefatigable neurotransmission of the sound. The molecular machinery ensuring the efficient, compensatory recycling of the synaptic vesicles (SVs), however, remains elusive. This study showed that hair cell knock-out of murine Dmxl2, whose human homolog is responsible for nonsyndromic sensorineural hearing loss DFNA71, resulted in auditory synaptopathy by impairing synaptic endocytosis and recycling. The mutant mice in the C57BL/6J background of either sex had mild hearing loss with severely diminished wave I amplitude of the auditory brainstem response. Membrane capacitance measurements of the IHCs revealed deficiency in sustained synaptic exocytosis and endocytic membrane retrieval. Consistent with the electrophysiological findings, 3D electron microscopy reconstruction showed reduced reserve pool of SVs and endocytic compartments, while the membrane-proximal and ribbon-associated vesicles remain intact. Our results propose an important role of DMXL2 in hair cell endocytosis and recycling of the SVs.
Sigma-Aldrich
Anti-GLUR2 antibody produced in rabbit, affinity isolated antibody