MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Chemical exchange saturation transfer by intermolecular double-quantum coherence.
Chemical exchange saturation transfer by intermolecular double-quantum coherence.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2008-06-24)
Wen Ling, Uzi Eliav, Gil Navon, Alexej Jerschow
ABSTRACT
A number of contrast enhancement effects based on the use of intermolecular multiple-quantum coherences, or distant dipolar field effects are known. This phenomenon is characterized by the dependence on the mth power of the initial magnetization (where m is the coherence order used). In this paper, we describe the contrast enhancement based on chemical exchange saturation transfer and NOE, which is achieved by the use of intermolecular double-quantum coherences (iDQC). The method was validated using clinically relevant systems based on glycosaminoglycans and a sample of cartilage tissue, showing that the CEST contrast, as well as, NOE are enhanced by iDQC.