MilliporeSigma
Cathepsin D: a cellular roadmap.
Biochemical and biophysical research communications (2008-09-03)
Nousheen Zaidi, Andreas Maurer, Sebastian Nieke, Hubert Kalbacher
ABSTRACT
Cathepsin D is a normal and major component of lysosomes, it is found in almost all cells and tissues of mammals. Present review describes different events in cellular life of cathepsin D mainly its biosynthesis, co-translational and posttranslational modifications, targeting to lysosomes and proteolytic processing and maturation within lysosomes.
Sigma-Aldrich
Cathepsin D human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)