MilliporeSigma
  • Doublecortin is a highly valuable endogenous marker of adult neurogenesis in canaries. Commentary on Vellema M et al. (2014): Evaluating the predictive value of doublecortin as a marker for adult neurogenesis in canaries (Serinus canaria) . J Comparative Neurol 522:1299-1315.
Doublecortin is a highly valuable endogenous marker of adult neurogenesis in canaries. Commentary on Vellema M et al. (2014): Evaluating the predictive value of doublecortin as a marker for adult neurogenesis in canaries (Serinus canaria) . J Comparative Neurol 522:1299-1315.
Brain, behavior and evolution (2014-07-19)
Jacques Balthazart, Gregory F Ball
ABSTRACT
Commentary on “Evaluating the Predictive Value of Doublecortin as a Marker for Adult Neurogenesis in Canaries (Serinus canaria)” by Michiel Vellema, Moritz Hertel, Susan L. Urbanus, Annemie Van der Linden, and Manfred Gahr (Journal of Comparative Neurology 2014, 522 : 1299-1315)
Sigma-Aldrich
Anti-Doublecortin Antibody, serum, from guinea pig