MilliporeSigma
Improved PIEZO1 agonism through 4-benzoic acid modification of Yoda1.
British journal of pharmacology (2022-12-03)
Gregory Parsonage, Kevin Cuthbertson, Naima Endesh, Nicoletta Murciano, Adam J Hyman, Charlotte H Revill, Oleksandr V Povstyan, Eulashini Chuntharpursat-Bon, Marjolaine Debant, Melanie J Ludlow, Timothy Simon Futers, Laeticia Lichtenstein, Jacob A Kinsella, Fiona Bartoli, Maria Giustina Rotordam, Nadine Becker, Andrea Brüggemann, Richard Foster, David J Beech
ABSTRACT
The protein PIEZO1 forms mechanically activated, calcium-permeable, non-selective cation channels in numerous cell types from several species. Options for pharmacological modulation are limited and so we modified a small-molecule agonist at PIEZO1 channels (Yoda1) to increase the ability to modulate these channels. Medicinal chemistry generated Yoda1 analogues that were tested in intracellular calcium and patch-clamp assays on cultured cells exogenously expressing human or mouse PIEZO1 or mouse PIEZO2. Physicochemical assays and wire myography assays on veins from mice with genetic disruption of PIEZO1. A Yoda1 analogue (KC159) containing 4-benzoic acid instead of the pyrazine of Yoda1 and its potassium salt (KC289) have equivalent or improved reliability, efficacy and potency, compared with Yoda1 in functional assays. Tested against overexpressed mouse PIEZO1 in calcium assays, the order of potency (as EC50 values, nM) was KC289, 150 > KC159 280 > Yoda1, 600). These compounds were selective for PIEZO1 over other membrane proteins, and the physicochemical properties were more suited to physiological conditions than those of Yoda1. The vasorelaxant effects were consistent with PIEZO1 agonism. In contrast, substitution with 2-benzoic acid failed to generate a modulator. 4-Benzoic acid modification of Yoda1 improves PIEZO1 agonist activity at PIEZO1 channels. We suggest naming this new modulator Yoda2. It should be a useful tool compound in physiological assays and facilitate efforts to identify a binding site. Such compounds may have therapeutic potential, for example, in diseases linked genetically to PIEZO1 such as lymphatic dysplasia.
Sigma-Aldrich
Yoda2, ≥98% (HPLC)