MilliporeSigma
  • Posttranslational splicing modifications as a key mechanism in cytarabine resistance in acute myeloid leukemia.
Posttranslational splicing modifications as a key mechanism in cytarabine resistance in acute myeloid leukemia.
Leukemia (2023-07-09)
María Luz Morales, Roberto García-Vicente, Alba Rodríguez-García, Armando Reyes-Palomares, África Vincelle-Nieto, Noemí Álvarez, Alejandra Ortiz-Ruiz, Vanesa Garrido-García, Alicia Giménez, Gonzalo Carreño-Tarragona, Ricardo Sánchez, Rosa Ayala, Joaquín Martínez-López, María Linares
ABSTRACT
Despite the approval of several drugs for AML, cytarabine is still widely used as a therapeutic approach. However, 85% of patients show resistance and only 10% overcome the disease. Using RNA-seq and phosphoproteomics, we show that RNA splicing and serine-arginine-rich (SR) proteins phosphorylation were altered during cytarabine resistance. Moreover, phosphorylation of SR proteins at diagnosis were significantly lower in responder than non-responder patients, pointing to their utility to predict response. These changes correlated with altered transcriptomic profiles of SR protein target genes. Notably, splicing inhibitors were therapeutically effective in treating sensitive and resistant AML cells as monotherapy or combination with other approved drugs. H3B-8800 and venetoclax combination showed the best efficacy in vitro, demonstrating synergistic effects in patient samples and no toxicity in healthy hematopoietic progenitors. Our results establish that RNA splicing inhibition, alone or combined with venetoclax, could be useful for the treatment of newly diagnosed or relapsed/refractory AML.
Sigma-Aldrich
Anti-SR protein family Antibody, clone 16H3, clone 16H3, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Phosphoepitope SR proteins Antibody, clone 1H4, clone 1H4, from mouse
Pricing and availability is not currently available.