MilliporeSigma
  • Bromelain and ficin proteolytic effects on gliadin cytotoxicity and expression of genes involved in cell-tight junctions in Caco-2 cells.
Bromelain and ficin proteolytic effects on gliadin cytotoxicity and expression of genes involved in cell-tight junctions in Caco-2 cells.
Amino acids (2023-10-07)
Masoumeh Sadat Mousavi Maleki, Hooman Aghamirza Moghim Ali Abadi, Behrooz Vaziri, Ali Akbar Shabani, Ghazaleh Ghavami, Hamid Madanchi, Soroush Sardari
ABSTRACT
Enzyme therapy for celiac disease (CeD), which digests gliadin into non-immunogenic and non-toxic peptides, can be an appropriate treatment option for CeD. Here, we have investigated the effectiveness of bromelain and ficin on gliadin digestion using in vitro, such as SDS-PAGE, HPLC, and circular dichroism (CD). Furthermore, the cytotoxicity of gliadin and 19-mer peptide before and after digestion with these enzymes was evaluated using the MTT assay in the Caco-2 cell line. Finally, we examined the effect of these treatments along with Larazotide Acetate on the expression of genes involved in cell-tight junctions, such as Occludin, Claudin 3, tight junction protein-1, and Zonulin in the Caco-2 cell line. Our study demonstrated bromelain and ficin digestion effects on the commercial and wheat-extracted gliadin by SDS-PAGE, HPLC, and CD. Also, the cytotoxicity results on Caco-2 showed that toxicity of the gliadin and synthetic 19-mer peptide was decreased by adding bromelain and ficin. Furthermore, the proteolytic effects of bromelain and ficin on gliadin indicated the expression of genes involved in cell-tight junctions was improved. This study confirms that bromelain and ficin mixture could be effective in improving the symptoms of CeD.
Sigma-Aldrich
Larazotide acetate salt, ≥95% (HPLC)