MilliporeSigma
  • Aryl hydrocarbon receptor sulfenylation promotes glycogenolysis and rescues cancer chemoresistance.
Aryl hydrocarbon receptor sulfenylation promotes glycogenolysis and rescues cancer chemoresistance.
The Journal of clinical investigation (2023-12-15)
Nannan Zhou, Jie Chen, Zheng Ling, Chaoqi Zhang, Yabo Zhou, Dianheng Wang, Li Zhou, Zhenfeng Wang, Nan Sun, Xin Wang, Huafeng Zhang, Ke Tang, Jingwei Ma, Jiadi Lv, Bo Huang
ABSTRACT
Elevation of reactive oxygen species (ROS) levels is a general consequence of tumor cells' response to treatment and may cause tumor cell death. Mechanisms by which tumor cells clear fatal ROS, thereby rescuing redox balance and entering a chemoresistant state, remain unclear. Here, we show that cysteine sulfenylation by ROS confers on aryl hydrocarbon receptor (AHR) the ability to dissociate from the heat shock protein 90 complex but to bind to the PPP1R3 family member PPP1R3C of the glycogen complex in drug-treated tumor cells, thus activating glycogen phosphorylase to initiate glycogenolysis and the subsequent pentose phosphate pathway, leading to NADPH production for ROS clearance and chemoresistance formation. We found that basic ROS levels were higher in chemoresistant cells than in chemosensitive cells, guaranteeing the rapid induction of AHR sulfenylation for the clearance of excess ROS. These findings reveal that AHR can act as an ROS sensor to mediate chemoresistance, thus providing a potential strategy to reverse chemoresistance in patients with cancer.
Sigma-Aldrich
Anti-Sulfenic Acid Modified Cysteine (2-Thiodimedone-Specific Ig) Antibody, serum, from rabbit
Pricing and availability is not currently available.