MilliporeSigma
  • Cyclic fasting bolsters cholesterol biosynthesis inhibitors' anticancer activity.
Cyclic fasting bolsters cholesterol biosynthesis inhibitors' anticancer activity.
Nature communications (2023-11-01)
Amr Khalifa, Ana Guijarro, Silvia Ravera, Nadia Bertola, Maria Pia Adorni, Bianca Papotti, Lizzia Raffaghello, Roberto Benelli, Pamela Becherini, Asmaa Namatalla, Daniela Verzola, Daniele Reverberi, Fiammetta Monacelli, Michele Cea, Livia Pisciotta, Franco Bernini, Irene Caffa, Alessio Nencioni
ABSTRACT
Identifying oncological applications for drugs that are already approved for other medical indications is considered a possible solution for the increasing costs of cancer treatment. Under the hypothesis that nutritional stress through fasting might enhance the antitumour properties of at least some non-oncological agents, by screening drug libraries, we find that cholesterol biosynthesis inhibitors (CBIs), including simvastatin, have increased activity against cancers of different histology under fasting conditions. We show fasting's ability to increase CBIs' antitumour effects to depend on the reduction in circulating insulin, insulin-like growth factor-1 and leptin, which blunts the expression of enzymes from the cholesterol biosynthesis pathway and enhances cholesterol efflux from cancer cells. Ultimately, low cholesterol levels through combined fasting and CBIs reduce AKT and STAT3 activity, oxidative phosphorylation and energy stores in the tumour. Our results support further studies of CBIs in combination with fasting-based dietary regimens in cancer treatment and highlight the value of fasting for drug repurposing in oncology.
Sigma-Aldrich
ID8 Mouse Ovarian Surface Epithelial Cell Line, ID8 mouse ovarian surface epithelial cell line is frequently used as a syngeneic mouse model for human ovarian cancer.