MilliporeSigma
Synthesis of a proposed thymic factor.
Journal of medicinal chemistry (1979-05-01)
R G Strachan, W J Paleveda, S J Bergstrand, R F Nutt, F W Holly, D F Veber
PMID458811
ABSTRACT
Less than Glu-Ala-Lys-Ser-Gln-Gly-Gly-Ser-Asn, a proposed serum thymic factor, has been synthesized. The protected precursor, less than Glu-Ala-Lys(i-Noc)-ser(Bzl)-Gln-Gly-Gly-Ser(Bzl)-Asn, was prepared by a combination of solid phase and solution methods. The benzyl blocking groups were removed by HF and the i-Noc blocking group was removed by catalytic hydrogenation.