MilliporeSigma
TIMP-2 and IGFBP7 in human kidney biopsies in renal disease.
Clinical kidney journal (2023-09-04)
Moritz Schanz, Martin Kimmel, Mark Dominik Alscher, Kerstin Amann, Christoph Daniel
ABSTRACT
Tissue inhibitor of matrix metalloproteinase-2 (TIMP-2) and insulin-like growth factor binding protein 7 (IGFBP7) are markers of tubular stress and urinary [TIMP-2]*[IGFBP7] is an established biomarker for risk assessment of acute kidney injury. There are no studies of expression profiles or localization of these markers in human renal tissue with confirmed renal disease. We analysed 37 kidney biopsies of patients with renal disease and 10 non-diseased control biopsies for TIMP-2 and IGFBP7 expression using immunohistochemistry. Changes in glomerular morphology were evaluated by a semi-quantitative glomerulosclerosis score (GSI) and tubular interstitial changes were graded by the tubular injury score (TSI) using periodic acid-Schiff-stained paraffin sections. Interstitial fibrosis and tubular atrophy (IF/TA) were graded according to the Banff classification. Urinary [TIMP-2]*[IGFBP7] was collected at the time of biopsy. TIMP-2 and IGFBP7 had significantly greater expression in kidney biopsies from patients with renal disease compared with control tissue, especially in the tubular compartment. Here, IGFBP7 was detected in proximal and distal tubules while TIMP-2 was predominantly localized in the collecting ducts. Renal injury significantly correlated with staining intensity for TIMP-2 and IGFBP7: GSI weakly correlated with glomerular TIMP-2 (r = 0.36) and IGFBP7 (r = 0.35) and TSI correlated with tubular TIMP-2 (r = 0.41) and IGFBP7 (r = 0.43). Urinary [TIMP-2]*[IGFBP7] correlated weakly with the histopathological damage score but not with glomerular and tubular expression. Our findings underline the role of TIMP-2/IGFBP7 as an unspecific marker of renal injury that is already in use for early detection of acute kidney injury.
Sigma-Aldrich
Anti-AQP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution