MilliporeSigma
  • Zbtb16 mediates a switch between Fgf signalling regimes in the developing hindbrain.
Zbtb16 mediates a switch between Fgf signalling regimes in the developing hindbrain.
Development (Cambridge, England) (2023-08-29)
Sami A Leino, Sean C J Constable, Andrea Streit, David G Wilkinson
ABSTRACT
Developing tissues are sequentially patterned by extracellular signals that are turned on and off at specific times. In the zebrafish hindbrain, fibroblast growth factor (Fgf) signalling has different roles at different developmental stages: in the early hindbrain, transient Fgf3 and Fgf8 signalling from rhombomere 4 is required for correct segmentation, whereas later, neuronal Fgf20 expression confines neurogenesis to specific spatial domains within each rhombomere. How the switch between these two signalling regimes is coordinated is not known. We present evidence that the Zbtb16 transcription factor is required for this transition to happen in an orderly fashion. Zbtb16 expression is high in the early anterior hindbrain, then gradually upregulated posteriorly and confined to neural progenitors. In mutants lacking functional Zbtb16, fgf3 expression fails to be downregulated and persists until a late stage, resulting in excess and more widespread Fgf signalling during neurogenesis. Accordingly, the spatial pattern of neurogenesis is disrupted in Zbtb16 mutants. Our results reveal how the distinct stage-specific roles of Fgf signalling are coordinated in the zebrafish hindbrain.
Sigma-Aldrich
Tyramine hydrochloride, ≥98%
Sigma-Aldrich
SU-5402, ≥98% (HPLC)