MilliporeSigma
  • Correlation between Serum Myosin Light Chain 4 Levels and Recurrence after Radiofrequency Ablation in Patients with Atrial Fibrillation.
Correlation between Serum Myosin Light Chain 4 Levels and Recurrence after Radiofrequency Ablation in Patients with Atrial Fibrillation.
International heart journal (2023-07-31)
Yibo Yu, Renyuan Fang, He Jin, Binhao Wang, Fang Gao, Bin He
ABSTRACT
Atrial fibrillation (AF) is the most common arrhythmia that is harmful to human health. This study aims to explore the relationship between myosin light chain 4 (MYL4) and AF recurrence after radiofrequency ablation (RFA). Patients with AF (n = 85) were enrolled, and healthy subjects (n = 90) with normal sinus rhythm and no previous history of AF were selected as controls. The serum levels of MYL4, transforming growth factor (TGF) -β1, and procollagen type-I C-terminal propeptide (PICP) were determined. The correlation between MYL4 and atrial fibrosis remodeling indicators (TGF-β1/PICP) and left atrial diameter (LAD) was analyzed. The influence of MYL4 on AF recurrence after RFA was evaluated, and the independent correlation between them was assessed. Patients with AF and the controls showed no significant differences in age, gender, body mass index, systolic blood pressure, diastolic blood pressure, left ventricular ejection fraction, triglycerides, total cholesterol, high-density lipoprotein, low-density lipoprotein, white blood cell count, neutrophil/lymphocyte ratio, brain natriuretic peptide, and history of smoking, drinking, hypertension, and diabetes (P > 0.05), but with increased LAD in patients with AF (P < 0.01). Serum MYL4 level was reduced in patients with AF (0.6 ± 0.2) compared with that of controls (0.1 ± 0.6) (P < 0.01), and it was negatively correlated with TGF-β1, PICP, and LAD (r = -0.2389, P < 0.05; r = -0.5174, P < 0.01; r = -0.3191; P < 0.01). Low levels of MYL4 increased the risk of AF recurrence after RFA (χ2 = 16.64; P < 0.0001). A low MYL4 level in patients with AF showed a poorer prognosis. Serum MYL4 level and AF type were independent risk factors affecting AF recurrence after RFA.
Sigma-Aldrich
Anti-Procollagen Type I Antibody, CT, clone PCIDG10 (Ascites Free), clone PCIDG10, from mouse