MilliporeSigma
  • Coronin 2B Regulates Neuronal Migration via Rac1-Dependent Multipolar-Bipolar Transition.
Coronin 2B Regulates Neuronal Migration via Rac1-Dependent Multipolar-Bipolar Transition.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2023-01-15)
Yuewen Chen, Zhenyan Xu, Jing Chen, Yue Qiu, Lin Yuan, Peng Liu, Jing Duan, Li Chen, Yu Chen
ABSTRACT
In the developing cortex, excitatory neurons migrate along the radial fibers to their final destinations and build up synaptic connection with each other to form functional circuitry. The shaping of neuronal morphologies by actin cytoskeleton dynamics is crucial for neuronal migration. However, it is largely unknown how the distribution and assembly of the F-actin cytoskeleton are coordinated. In the present study, we found that an actin regulatory protein, coronin 2B, is indispensable for the transition from a multipolar to bipolar morphology during neuronal migration in ICR mice of either sex. Loss of coronin 2B led to heterotopic accumulation of migrating neurons in the intermediate zone along with reduced dendritic complexity and aberrant neuronal activity in the cortical plate. This was accompanied by increased seizure susceptibility, suggesting the malfunction of cortical development in coronin 2B-deficient brains. Coronin 2B knockdown disrupted the distribution of the F-actin cytoskeleton at the leading processes, while the migration defect in coronin 2B-deficient neurons was partially rescued by overexpression of Rac1 and its downstream actin-severing protein, cofilin. Our results collectively reveal the physiological function of coronin 2B during neuronal migration whereby it maintains the proper distribution of activated Rac1 and the F-actin cytoskeleton.SIGNIFICANCE STATEMENT Deficits in neuronal migration during cortical development result in various neurodevelopmental disorders (e.g., focal cortical dysplasia, periventricular heterotopia, epilepsy, etc.). Most signaling pathways that control neuronal migration process converge to regulate actin cytoskeleton dynamics. Therefore, it is important to understand how actin dynamics is coordinated in the critical processes of neuronal migration. Herein, we report that coronin 2B is a key protein that regulates neuronal migration through its ability to control the distribution of the actin cytoskeleton and its regulatory signaling protein Rac1 during the multipolar-bipolar transition in the intermediate zone, providing insights into the molecular machinery that drives the migration process of newborn neurons.
Sigma-Aldrich
Anti-CORO2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution