MilliporeSigma
  • Reduced membrane cholesterol content in skeletal muscle is not essential for greater insulin-stimulated glucose uptake after acute exercise by rats.
Reduced membrane cholesterol content in skeletal muscle is not essential for greater insulin-stimulated glucose uptake after acute exercise by rats.
Applied physiology, nutrition, and metabolism = Physiologie appliquee, nutrition et metabolisme (2021-03-26)
Haiyan Wang, Edward B Arias, Gregory D Cartee
ABSTRACT
One exercise session can elevate insulin-stimulated glucose uptake (GU) by skeletal muscle, but it is uncertain if this effect is accompanied by altered membrane cholesterol content, which is reportedly inversely related to insulin-stimulated GU. Muscles from sedentary (SED) or exercised 3 h post-exercise (3hPEX) rats were evaluated for GU, membrane cholesterol, and phosphorylation of cholesterol regulatory proteins (pHMCGRSer872 and pABCA1Ser2054). Insulin-stimulated GU for 3hPEX exceeded SED. Membrane cholesterol, pHMCGRSer872 and pABCA1Ser2054 did not differ between groups. Novelty: Alterations in membrane cholesterol and phosphorylation of proteins that regulate muscle cholesterol are not essential for elevated insulin-stimulated GU in skeletal muscle after acute exercise.
Sigma-Aldrich
Anti-AS160 Antibody, from rabbit, purified by affinity chromatography