MilliporeSigma
  • Overexpression of heat-shock protein 47 impacts survival of patients with oral squamous cell carcinoma.
Overexpression of heat-shock protein 47 impacts survival of patients with oral squamous cell carcinoma.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology (2023-05-29)
Bruno Cesar da Costa, Mauricio Rocha Dourado, Everton Freitas de Moraes, Luana Marí Panini, Amr Elseragy, Fábio Haach Téo, Gustavo Narvaes Guimarães, Renato Assis Machado, Maija Risteli, Clarissa Araujo Gurgel Rocha, Lívia Máris Ribeiro Paranaíba, Wilfredo Alejandro González-Arriagada, Sabrina Daniela da Silva, Ana Lucia Carrinho Ayroza Rangel, Marcelo Rocha Marques, Tuula Salo, Ricardo D Coletta
ABSTRACT
The expression of heat-shock protein 47 (HSP47) has been linked to collagen synthesis control and implicated in fibrotic disorders, but more recent studies have demonstrated its role in solid tumors. In this study, we explored the prognostic impact of HSP47 in oral squamous cell carcinomas (OSCC) and determined the in vitro effects of its loss-of-function on viability, proliferation, migration, invasion, and resistance to cisplatin of OSCC cells. The HSP47 expression in tumor samples was assessed by immunohistochemistry in two independent cohorts totaling 339 patients with OSCC, and protein levels were associated with clinicopathological features and survival outcomes. The OSCC cell lines HSC3 and SCC9 were transduced with lentivirus expressing short hairpin RNA to stably silence HSP47 and used in assays to measure cellular viability, proliferation, migration, and invasion. HSP47 was overexpressed in OSCC samples, and its overexpression was significantly and independently associated with poor disease-specific survival and shortened disease-free survival in both OSCC cohorts. The knockdown of HSP47 showed no effects on cell viability or cisplatin sensitivity, but impaired significantly proliferation, migration, and invasion of OSCC cells, with stronger effects on SCC9 cells. Our results show a significant prognostic impact of HSP47 overexpression in OSCC and reveal that HSP47 inhibition impairs the proliferation, migration, and invasion of OSCC cells. HSP47 may represent a potential therapeutic target for OSCC.
Sigma-Aldrich
Anti-SERPINH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution