MilliporeSigma
  • Matrix Stiffness Activating YAP/TEAD1-Cyclin B1 in Nucleus Pulposus Cells Promotes Intervertebral Disc Degeneration.
Matrix Stiffness Activating YAP/TEAD1-Cyclin B1 in Nucleus Pulposus Cells Promotes Intervertebral Disc Degeneration.
Aging and disease (2023-05-17)
Zijie Zhou, Yinxuan Suo, Jinyu Bai, Fanguo Lin, Xiang Gao, Huajian Shan, Yichao Ni, Xiaozhong Zhou, Lei Sheng, Jun Dai
ABSTRACT
Intervertebral disc degeneration is a leading cause of disability in the elderly population. Rigid extracellular matrix is a critical pathological feature of disc degeneration, leading to aberrant nucleus pulposus cells (NPCs) proliferation. However, the underlying mechanism is unclear. Here, we hypothesize that increased matrix stiffness induces proliferation and thus degenerative phenotypes of NPCs through YAP/TEAD1 signaling pathway. We established hydrogel substrates to mimic stiffness of degenerated human nucleus pulposus tissues. RNA-sequencing identified differentially expressed genes between primary rat NPCs cultured on rigid and soft hydrogels. Dual luciferase assay and gain- and loss-function experiments evaluated the correlation between YAP/TEAD1 and Cyclin B1. Furthermore, single-cell RNA-sequencing of human NPCs was performed to identify specific cell clusters with high YAP expression. Matrix stiffness increased in severely degenerated human nucleus pulposus tissues (p < 0.05). Rigid substrate enhanced rat NPCs proliferation mainly through Cyclin B1, which was directly targeted and positively regulated by YAP/TEAD1. Depletion of YAP or Cyclin B1 arrested G2/M phase progression of rat NPCs and reduced fibrotic phenotypes including MMP13 and CTGF (p < 0.05). Fibro NPCs with high YAP expression were identified in human tissues and responsible for fibrogenesis during degeneration. Furthermore, inhibition of YAP/TEAD interaction by verteporfin suppressed cell proliferation and alleviated degeneration in the disc needle puncture model (p < 0.05). Our results demonstrate that elevated matrix stiffness stimulates fibro NPCs proliferation through YAP/TEAD1-Cyclin B1 axis, indicating a therapeutic target for disc degeneration.
Sigma-Aldrich
Collagenase Type II, Cls II