MilliporeSigma
  • eIF3a regulation of mTOR signaling and translational control via HuR in cellular response to DNA damage.
eIF3a regulation of mTOR signaling and translational control via HuR in cellular response to DNA damage.
Oncogene (2022-03-14)
Shijie Ma, Zizheng Dong, Yanfei Huang, Jing-Yuan Liu, Jian-Ting Zhang
ABSTRACT
eIF3a (eukaryotic translation initiation factor 3a), a subunit of the eIF3 complex, has been suggested to play a regulatory role in protein synthesis and in cellular response to DNA-damaging treatments. S6K1 is an effector and a mediator of mTOR complex 1 (mTORC1) in regulating protein synthesis and integrating diverse signals into control of cell growth and response to stress. Here, we show that eIF3a regulates S6K1 activity by inhibiting mTORC1 kinase via regulating Raptor synthesis. The regulation of Raptor synthesis is via eIF3a interaction with HuR (human antigen R) and binding of the eIF3a-HuR complex to the 5'-UTR of Raptor mRNA. Furthermore, mTORC1 may mediate eIF3a function in cellular response to cisplatin by regulating synthesis of NER proteins and NER activity. Taken together, we conclude that the mTOR signaling pathway may also be regulated by translational control and mediate eIF3a regulation of cancer cell response to cisplatin by regulating NER protein synthesis.
Sigma-Aldrich
Anti-DEPTOR Antibody, from rabbit, purified by affinity chromatography