MilliporeSigma
  • Normal lymphocyte homeostasis and function in MALT1 protease-resistant HOIL-1 knock-in mice.
Normal lymphocyte homeostasis and function in MALT1 protease-resistant HOIL-1 knock-in mice.
The FEBS journal (2022-12-09)
Ioannis Skordos, Yasmine Driege, Mira Haegman, Marja Kreike, Jens Staal, Annelies Demeyer, Rudi Beyaert
ABSTRACT
The uniqueness of MALT1 protease activity in controlling several aspects of immunity in humans has made it a very attractive therapeutic target for multiple autoimmune diseases and lymphoid malignancies. Despite several encouraging preclinical studies with MALT1 inhibitors, severe reduction in regulatory T cells and immune-mediated pathology seen in MALT1 protease-dead (MALT1-PD) mice and some, but not all, studies analysing the effect of prolonged pharmacological MALT1 protease inhibition, indicates the need to further unravel the mechanism of MALT1 protease function. Notably, the contribution of individual MALT1 substrates to the immune defects seen in MALT1-PD mice is still unclear. Previous in vitro studies indicated a role for MALT1-mediated cleavage of the E3 ubiquitin ligase HOIL-1 in the modulation of nuclear factor-κB (NF-κB) signalling and inflammatory gene expression in lymphocytes. Here, we addressed the immunological consequences of inhibition of HOIL-1 cleavage by generating and immunophenotyping MALT1 cleavage-resistant HOIL-1 knock-in (KI) mice. HOIL-1 KI mice appear healthy and have no overt phenotype. NF-κB activation in T or B cells, as well as IL-2 production and in vitro T-cell proliferation, is comparable between control and HOIL-1 KI cells. Inhibition of HOIL-1 cleavage in mice has no effect on thymic T-cell development and conventional T-cell homeostasis. Likewise, B-cell development and humoral immune responses are not affected. Together, these data exclude an important role of MALT1-mediated HOIL-1 cleavage in T- and B-cell development and function in mice.
Sigma-Aldrich
Anti-RBCK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.