MilliporeSigma
  • Discovery and pharmacological characterization of N-[2-({2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl}amino)-2-methylpropyl]-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxamide hydrochloride (anagliptin hydrochloride salt) as a potent and selective DPP-IV inhibitor.
Discovery and pharmacological characterization of N-[2-({2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl}amino)-2-methylpropyl]-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxamide hydrochloride (anagliptin hydrochloride salt) as a potent and selective DPP-IV inhibitor.
Bioorganic & medicinal chemistry (2011-10-25)
Noriyasu Kato, Mitsuru Oka, Takayo Murase, Masahiro Yoshida, Masao Sakairi, Satoko Yamashita, Yoshika Yasuda, Aya Yoshikawa, Yuuji Hayashi, Mitsuhiro Makino, Motohiro Takeda, Yakufu Mirensha, Takuji Kakigami
ABSTRACT
In the course of our program for discovery of novel DPP-IV inhibitors, a series of pyrazolo[1,5-a]pyrimidines were found to be novel DPP-IV inhibitors. We identified N-[2-({2-[(2S)-2-cyanopyrrolidin-1-yl]-2-oxoethyl}amino)-2-methylpropyl]-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxamide hydrochloride (4a) and described its pharmacological profiles.
Sigma-Aldrich
Anagliptin, ≥98% (HPLC)
Pricing and availability is not currently available.