MilliporeSigma
  • Heparin-functionalized polymeric biomaterials in tissue engineering and drug delivery applications.
Heparin-functionalized polymeric biomaterials in tissue engineering and drug delivery applications.
Acta biomaterialia (2013-08-06)
Yingkai Liang, Kristi L Kiick
ABSTRACT
Heparin plays an important role in many biological processes via its interaction with various proteins, and hydrogels and nanoparticles comprising heparin exhibit attractive properties, such as anticoagulant activity, growth factor binding, and antiangiogenic and apoptotic effects, making them great candidates for emerging applications. Accordingly, this review summarizes recent efforts in the preparation of heparin-based hydrogels and formation of nanoparticles, as well as the characterization of their properties and applications. The challenges and future perspectives for heparin-based materials are also discussed. Prospects are promising for heparin-containing polymeric biomaterials in diverse applications ranging from cell carriers for promoting cell differentiation to nanoparticle therapeutics for cancer treatment.
Sigma-Aldrich
TissueFab® bioink , (GelHAHep)MA Vis/405 nm, low endotoxin
Pricing and availability is not currently available.
Sigma-Aldrich
TissueFab® bioink , (GelHep)MA Vis/ 405nm, low endotoxin
Pricing and availability is not currently available.