MilliporeSigma
  • Carbonic anhydrase inhibitors. Interaction of the antiepileptic drug sulthiame with twelve mammalian isoforms: kinetic and X-ray crystallographic studies.
Carbonic anhydrase inhibitors. Interaction of the antiepileptic drug sulthiame with twelve mammalian isoforms: kinetic and X-ray crystallographic studies.
Bioorganic & medicinal chemistry letters (2007-06-26)
Claudia Temperini, Alessio Innocenti, Antonio Mastrolorenzo, Andrea Scozzafava, Claudiu T Supuran
ABSTRACT
Sulthiame, a clinically used antiepileptic, was investigated for its interaction with 12 catalytically active mammalian carbonic anhydrase (CA, EC 4.2.1.1) isoforms. The drug is a potent inhibitor of CA II, VII, IX, and XII (K(I)s of 6-56 nM), and a medium potency inhibitor against CA IV, VA, VB, and VI (K(I)s of 81-134 nM). The high resolution crystal structure of the hCA II-sulthiame adduct revealed a large number of favorable interactions between the drug and the enzyme which explain its strong low nanomolar affinity for this isoform and may also be exploited for the design of effective inhibitors incorporating sultam moieties.
Sigma-Aldrich
Sulthiame, ≥95% (HPLC)